X Drive Belt
Issue
Customer had an ASX-260 that would not move in the X direction
Environment
- ASX-260
- ASX-280
- ASX-520
- ASX-560
Resolution
- Even though the ASX-260 is discontinued, the 260/280/520/560 all use the same x drive belt.
- The part number for the X-drive belt is BRE0004481