Cetac Autosampler New Rinse Pump Clamp
Description:
New part number for the Rinse pump clamp on the ASX-560 and ASX-280
P/N: BRE0062617 Tubing Clip for DP2/DP3 Pumps on ASX autosampler
Environment:
- ASX-560
- ASX-280
New part number for the Rinse pump clamp on the ASX-560 and ASX-280
P/N: BRE0062617 Tubing Clip for DP2/DP3 Pumps on ASX autosampler