Chiller issues
Issue
Chiller Leaks
Environment
TF-900, TF-2500
Resolution
Re-installed stainless mesh in effluent opening.
Analysis
Cause
Stainless mesh fell into the reservoir allowing the fluid to spill over the top of the opening.
Chiller Leaks
TF-900, TF-2500
Re-installed stainless mesh in effluent opening.
Stainless mesh fell into the reservoir allowing the fluid to spill over the top of the opening.