ThermoFlex Error Codes
The Thermo Scientific ThermoFlex 2-11 chiller displays error codes to indicate issues. If the chiller is still running, press enter to see if the code clears. If the chiller shut down, the error code will flash until addressed. Common error codes include communication errors, low fluid levels, and high/low temperature or pressure. Corrective actions involve checking connections, adjusting settings, cleaning filters, and ensuring proper ventilation. Restart the chiller after resolving the issue.
Description:
This document lists the error codes encountered on the ThermoFlex line of chillers, possible causes and potential actions which can be taken to resolve the issue.
Environment:
- ThermoFlex 900
- ThermoFlex 2500
Attachment(s):
| File | Last Modified |
|---|---|
| ThermoFlex Error Codesl.pdf | December 21, 2022 |