Thermoflex Chiller Error 41
Issue
The Thermoflex chiller displays an Error 41 causing the chiller to stop.
Environment
- Thermoflex 900
- Thermoflex 1400
- Thermoflex 2500
Resolution
1. Verify that the display cable is properly attached (please see Attachments "ThermoFlex Error 41 Checks").
2. If problem persists replace the display panel. Contact Thermoflex support at 800-258-0830 (option 2) to find the correct part number using the serial number of the chiller.
2a. Remove the right and left side panels by removing the two screws securing the cover to the unit.
2b. Undo the connection
2c. Remove the two screws securing the panel to the unit.
2d. Remove the two remaining screws and the 5/16" nut
2e. Undo the ribbon cable and the four screws securing the board to the panel. NOTE: the orientation of the ribbon cable, ensure it does not get twisted when installing the replacement board.
Cause
The Thermoflex chiller cannot communicate with the display due to a problem with the display cable connection or the display panel.
Attachment(s)
File | Last Modified |
---|---|
clipboard_e2e2b119aee2608095f998417313ca506.png | January 11, 2023 |
ThermoFlex Error 41 Checks.ppt | January 11, 2023 |
clipboard_ef60027419b4ccdd9af33661544c82256.png | January 11, 2023 |
clipboard_e54c281493de66c5077f05ee74af10648.png | January 11, 2023 |
clipboard_e04f2b6d0411cfa17e7558aba6424b8e1.png | January 11, 2023 |
clipboard_ebfed2bff8aa83fa6ab96bfb007e56658.png | January 11, 2023 |
clipboard_e9f129fc56e8b49eb8a76f3415ba0496a.png | January 11, 2023 |
clipboard_ec7d2d91cbf243d9bdd002bf587727a3b.png | January 11, 2023 |
P1.jpg | January 11, 2023 |