Default probe for non-FAST autosamplers
Description
The default probe used with non-FAST autosamplers is a 0.5mm ID probe (PN 1301750) identified with the orange maker.
Environment
ESI 2/4/8/16 SC-DX autosamplers withou FAST or PrepFAST
The default probe used with non-FAST autosamplers is a 0.5mm ID probe (PN 1301750) identified with the orange maker.
ESI 2/4/8/16 SC-DX autosamplers withou FAST or PrepFAST