Probe not going to rinse after Qtegra update
Issue
ESI autosampler probe not washing between samples after updating from Qtegra 2.10 to Qtegra 2.18SR1.
Environment
- Qtegra
- SC-2/4/8 DX
Resolution
Remove the FAST method from the lab book and run as normal.
Cause
In the newer versions of Qtegra and the ESI plugin, there is no need for a dummy FAST method when bypassing the FAST valve.