Average of Multiple Samples in Qtegra
Issue
How do you calculate the average of multiple samples in Qtegra in order to satisfy some methodologies, for example System Suitability test for USP
Environment
Qtegra
Resolution
1) Save the Qtegra report file Average of Multiple Samples.qrpt to the following directory: C:\C:\ProgramData\Thermo\Qtegra\_Application Data\Reporting
2) Restart the Computer
3) While in the LabBook navigate to Reports and then click on the “Average of Multiple Samples” report
4) Once the report has generated, click on “Edit Report”
5) Select “Result Table:Result Data Table” in the Report content menu
6) Navigate to Conditions and then in the Condition drop down menu select the samples you would like to be averaged.
7) Click the Execute Report button to generate the report
At this point you can export the report as a PDF or print a hard copy
Note: you must complete steps 1-4 each time you want to generate a report of the average of a different set of samples.
Analysis
Cause
Attachment(s)
File | Last Modified |
---|---|
Execute Report.png | April 05, 2022 |
Conditions.png | April 05, 2022 |
Result Data Table.png | April 05, 2022 |
Edit Report.png | April 05, 2022 |
Avg of Multiple Samples report.png | April 05, 2022 |
Average of Multiple Samples.qrpt | April 05, 2022 |