Thermo Fisher Scientific
Determining the Autosampler COM Port
Issue
How to determine the COM port the autosampler is communicating through
Environment
  • Cetac
  • ESI
  • Qtegra
Resolution
Open Device Manager within Windows
Device Manager.png
Expand the tree for Ports by left-clicking on the triangle next to Ports
Ports.png
Look for the autosampler COM port. In the example below, the autosampler is set up on COM5
AS Com Port.png
  • Was this article helpful?