Thermo Fisher Scientific
Displaying the ProgramData Folder in Windows 10
Description: 
1)    While in the File Explorer, click on the View tab
2)    Uncheck the checkbox for Hidden items
3)    The ProgramData folder will now be visible when navigating to the C: drive. The transparent icon indicates it is a Hidden folder that is being displayed.
Environment:
  • iCAP Pro
  • iCAP 7000 Series
  • iCAP Q
  • iCAP Qnova Series
  • Was this article helpful?