How to Enable Quality Control Tab
Issue
Quality Control Tab in Method Parameters is not there
Environment
- Qtegra
Resolution
- Under method parameters choose the Quantification tab
- Check the "Use Quality Control" box
- The Quality Control tab is now present under Ratios