Importing Lab Books into Qtegra
Issue
Importing LabBooks into Qtegra
Environment
- Qtegra
Resolution
- Open Qtegra
- In the left hand panel select LabBooks
- Towards the bottom int Open LabBook section click open
- In the left hand panel select LabBooks and then right click on LabBooks
- In the pop up box select import
- Use the file manager to locate the LabBook you would like to import.
- The LabBook should now be visable and able to use.