Thermo Fisher Scientific
Output Sample Results in Different Units Than Defined Standards
This article explains how to configure sample results to display in a different unit than the units defined for standards in Qtegra. Use the Add Customized Trace option within Quantification to assign alternate reporting units for one or multiple analytes.
Issue
Sample results need to display in units different from the configured standard units in Qtegra.
Environment
  • iCAP Qnova Series
  • iCAP 7000 Series
  • iCAP PRO Series
Resolution
Add a Customized Trace
  1. Click Quantification.
  2. Right-click the analyte row and select Add customized trace.
    Add customized trace.png
    Figure 1. Add Customized Trace option within Quantification
  3. To modify multiple analytes simultaneously, hold Ctrl while selecting analytes, then right-click and select Add customized trace.
  4. From the Unit dropdown menu, select the desired reporting unit.
    Select Unit for Customized Trace.png
    Figure 2.  Select Desired Reporting Units
  5. Verify the displayed sample results update to the selected unit.