Thermo Fisher Scientific
Performance Report is blank
Issue
Error in log file indicates that PerformanceReport.qrpt file is missing in folder: C:\ProgramData\Thermo\Qtegra\_Application Data\PluginData\BrigidMS\Model\iCAPRQ\Reports
Note: the exact file path will vary depending on what type of instrument Qtegra is controlling
Copied PerformanceReport.qrpt from backed up location which resolved the error. Modified performance report to include Ba++ and adjusted limits for Ce++ ratio as well. Tested performance report generation and no error occurred
Environment
  • iCAP Qnova Series
Resolution
Copied PerformanceReport.qrpt from backed up location which resolved the error.
  • Was this article helpful?