Qtegra configuration has been corrupted and Qtegra fails to open
Issue:
Qtegra configuration has been corrupted and Qtegra fails to open
Environment:
Qtegra
Resolution:
- Close all Qtegra applications.
- Delete the following folder: C:\Users\user-name\AppData\Local\Thermo_Fisher_Scientific\Qtegra.exe_StrongName_f4vvpfoqywl5qcnhqsiczgblqzkvsmp0.
- Restart Qtegra.