Qtegra for ICP-OES and ICP-MS Shutting Down the Plasma Automatically Upon LabBook Completion
Description:
1) Within the Scheduler, click on Settings
2) “Closedown” aka Automatic Plasma Shutdown defaults to “Never Closedown”. Please select a different option from the dropdown menu. For instance, if you select “Closedown on an empty queue”, the plasma will shut off once the Scheduler is empty (there are no more LabBooks to be ran).
Environment:
Qtegra
Attachment(s):
File | Last Modified |
---|---|
Shutdown the Plasma Automatically.png | February 21, 2023 |
Scheduler Settings.png | February 21, 2023 |
Qtegra for ICP-OES and ICP-MS Shutting Down the Plasma Automatically Upon LabBook Completion.pdf | July 29, 2022 |