Restart Qtegra Services to Restore Instrument Control or LabBook Updates
Restart Qtegra Windows services to resolve issues where LabBook progress stops updating or instrument control is lost.
Issue
• LabBook progress does not update during scheduled runs
• Loss of instrument control
Environment
• Qtegra ISDS Software
• Windows Operating System
• Qtegra (2.10 SR7 and later for Post Process Service)
• Applicable connected instruments
Resolution
Prerequisite:
Close Qtegra, Configurator, and all related Qtegra applications before restarting services.
  1. Click on the Search icon.png  button if available in Windows taskbar or Right-click on Windows icon and select Search
  2. Type "Services" into the Windows search box and select Run as administrator.
    Services.png
  3.  Click on Description Column Header.png to sort the Services by Description
  4.  Scroll down to the list Qtegra services
    Qtegra Services List.png
  5. Right-click on the Logging Service and select Stop
    Stop Logging Service.png
  6. A Stop Other Services popup window will appear. Click Yes to stop the other Qtegra Services
    Stop Other Services.png
  7. Right-click on Acquisition Service and select Start
    Start Acquisition Service.png
  8. The Post Process Service was added in Qtegra 2.10 SR7 (2.10.4345.39). For Qtegra 2.10 SR7 or later: Right-click the Post Process Service and select Start.
    Start Post Process Service.png
  9.  Once all of the Qtegra services have been restarted, Running should be displayed in the Status column for all of the Qtegra services.
    Qtegra Services Running.png
    Expected Result:
    All Qtegra services display "Running" in the Status column.
  10. Restart Qtegra
NOTE: If this does not work, please contact technical support for further assistance.