Restarting Qtegra Services
Issue
Issues such as those bulleted below will occur as the result of one or more of the Qtegra services no longer working properly.
- Progress of LabBook which has been scheduled to run is not updating
- No control of instrument
Environment
Qtegra
Resolution
Prerequisite:
Close out of Qtegra, Configurator and any other related Qtegra applications
1) Click on the button if available in Windows taskbar or Right-click on Windows icon and select Search
2) Type "Services" into the search box and then click "Run as Administrator"
3) Click on to sort the Services by Description
4) Scroll down to the list Qtegra services
5) Right-click on the Logging Service and select Stop
6) A Stop Other Services popup window will appear. Click Yes to stop the other Qtegra Services
7) Right-click on Acquisition Service and select Start
8) The Post Process Service was added in Qtegra 2.10 SR7 (2.10.4345.39). If the present version of Qtegra is 2.10 SR7 or later, , right-click on Post Process Service and select Start
9) Once all of the Qtegra services have been restarted, Running should be displayed in the Status column for all of the Qtegra services.
Analysis
Cause
Attachment(s)
File | Last Modified |
---|---|
Qtegra Services Running.png | July 20, 2022 |
Start Post Process Service.png | July 20, 2022 |
Start Acquisition Service.png | July 20, 2022 |
Stop Other Services.png | July 20, 2022 |
Stop Logging Service.png | July 20, 2022 |
Qtegra Services List.png | July 20, 2022 |
Services.png | July 20, 2022 |
Description Column Header.png | July 20, 2022 |
Search icon.png | July 20, 2022 |