Thermo Fisher Scientific
Turning on Automatic backup after analysis
Issue
Backing up Qtegra lab books automatically
Environment
  • Qtegra
Resolution
  1. Open Configurator Configurator icon.PNG
  2. Choose Settingsclipboard_e81549bd038e3d70a220142e97d8d5f5f.png
  3. Find and click Automatic backup after analysis
  4. Set the value on the right hand of the screen to True
    clipboard_ed8e55046756055a65b3fc37d6af036d2.png
Analysis
This will create a backup folder, found here: C:\ProgramData\Thermo\Qtegra\_Application Data\Workspace, that should still be backed up in a separate location for safe keeping.
  • Was this article helpful?