Thermo Fisher Scientific
Using Access Controls in Qtegra
Description: 
The Access Control Editor applet within Configurator allows for permissions to be set for access to the different programs and applications of Qtegra as well as specific functions within Qtegra. With the Access Control Editor, once can define who has access to programs or parts of programs and what type of access permission is granted or denied to a user or user group.
Environment:
Qtegra
  • Was this article helpful?