Sending a LabBook to Thermo Fisher
Issue
Need to send a LabBook to a Thermo Fisher Representative
Environment
- iCAP 6000
- iCAP 7000
- iCAP Q
- iCAP RQ
- Qtegra
Resolution
Get the Qtegra LabBook file to attach to an email by following the steps below
- In Qtegra, navigate to File Manager
- Right-click on the LabBook you would like to send and click Export
- Select a destination for the exported LabBook
- Email the LabBook as an attachment to the appropriate Thermo Fisher representative
Attachment(s)
File | Last Modified |
---|---|
destination.PNG | March 29, 2022 |
export.PNG | March 29, 2022 |
Qtegra File explorer ss.PNG | March 29, 2022 |