Qtegra Releases and Their Corresponding Version Number
Description:
Release Name |
Version |
Comments |
Release Date |
npQuant |
ESI |
Cetac |
2.2 |
2.2.1465.24 |
|
22.11.2013 |
|
|
|
2.2 SP1 |
2.2.1465.44 |
|
11.04.2014 |
|
|
|
2.2 SP2 |
2.2.1465.45 |
|
25.04.2014 |
|
|
|
2.4 |
2.4.1800.33 |
|
27.10.2014 |
|
2.6.8.7 |
3.0.0.2 |
2.4 SP1 |
2.4.1800.96 |
|
23.12.2014 |
|
2.6.8.9 |
3.0.0.2 |
2.4 SP2 |
2.4.1800.192 |
|
08.06.2015 |
|
2.6.8.21 |
3.0.0.2 |
2.6 |
2.6.2205.66 |
|
19.10.2015 |
|
2.6.8.21 |
3.0.0.2 |
2.6 SP1 |
2.6.2270.32 |
|
01.12.2015 |
|
2.6.8.21 |
3.0.0.2 |
2.6 SP2 |
2.6.2270.44 |
|
08.02.2016 |
|
2.6.8.21 |
3.0.0.2 |
2.7 |
2.7.2425.65 |
|
13.04.2016 |
1.2.2425.65 |
2.6.8.21 |
4.0.0.0 |
2.7 SR1 |
2.7.2425.154 |
|
15.08.2016 |
1.2.2425.154 |
2.6.8.21 |
4.1.1.0 |
2.7 SR1 |
2.7.2425.154 |
Re-release with updated Cetac plugin |
12.10.2016 |
1.2.2425.154 |
2.6.8.21 |
4.1.2.2 |
2.7 SR2 |
2.7.2426.165 |
(ICP-MS Only) |
14.12.2016 |
N/A |
2.6.8.21 |
4.1.2.2 |
2.8 |
2.8.2944.115 |
|
11.04.2017 |
1.3.2944.115 |
2.6.8.70 |
4.1.4.1 |
2.8 SR1 |
2.8.2944.202 |
|
30.06.2017 |
1.3.2944.202 |
2.6.8.70 |
4.1.4.1 |
2.8 SR2 |
2.8.3170.309 |
(ICP-MS Only) |
27.11.2017 |
1.3.3170.309 |
2.6.8.70 |
4.1.4.1 |
2.8 SR3 |
2.8.3170.392 |
|
24.01.2018 |
1.3.3170.392 |
2.6.8.70 |
4.1.4.1 |
2.10 |
2.10.3324.62 |
|
12.06.2018 |
1.4.3324.62 |
2.6.8.70 |
4.1.4.1 |
2.10 SR1 |
2.10.3324.83 |
|
17.08.2018 |
1.4.3324.83 |
2.6.8.109 |
4.1.4.1 |
2.10 SR2 |
2.10.3324.93 |
|
19.10.2018 |
|
|
|
2.10 SR3 |
2.10.3324.109 |
|
04.03.2019 |
|
|
|
2.10 SR4 |
2.10.3324.124 |
|
29.03.2019 |
|
|
|
2.10 SR5 |
2.10.3324.131 |
Last version to support Win 7 |
28.06.2019 |
1.4.3324.131 |
|
|
2.10 SR7 |
2.10.4345.39 |
|
30.01.2020 |
1.4.4345.39 |
2.6.8.109 |
4.1.4.1 |
2.10 SR8 |
2.10.4345.61 |
|
20.04.2020 |
|
|
|
2.10 SR8.1 |
2.10.4345.64 |
|
05.06.2020 |
|
2.6.8.109 |
4.3.2 |
2.10 SR9 |
2.10.4345.136 |
|
11.12.2020 |
1.4.4345.136 |
2.6.8.109 |
4.3.2 |
2.10 SR10 |
2.10.4345.236 |
Last version to support iCAP 6000/7000 Series |
June 2021 |
1.5.4345.218 |
2.6.10.89 |
4.4.0.3 |
2.14 |
2.14.5122.158 |
Only compatible with Windows 10 version 1607 or above |
Jan 2022 |
Not Supported |
2.6.10.89 |
4.4.0.4 |
2.14 SR1 |
2.14.5122.306 |
Win 10 version 1607 or above |
June 2022 |
Not supported |
2.6.10.89 |
4.4.0.4 |
NOTE: Qtegra 2.14 was the first version of Qtegra to support the iCAP PRO in conjunction with the iCAP Q/RQ/TQ.
iCAP PRO ONLY |
|
|
Release Name |
Version |
Comments |
2.11 |
2.11.4206.71 |
|
2.11 |
2.11.4206.302 |
|
2.11 |
2.11.4206.360 |
|
2.11 |
2.11.4206.441 |
|
2.11 SR1 |
2.11.4207.18 |
|
2.11 SR2 |
2.11.4776.211 |
|
Environment:
Qtegra