v 5.8 - 849940020021 - iTEVA Security Software Installation Manual
To install and configure iTEVA Security Software version 5.8, configure Windows Administration by adding iTEVA users to the Power Users group and restricting time/date changes to administrators. Install the Thermo Security Server and Administration Software on an NTFS-formatted disk. Configure security settings for the iTEVA Security client application. Install and configure SQL Server Express, create user logins, and install the iTEVA Security client application.
Description:
This manual explains how to install and configure the Thermo Scientific iTEVA Security software
Environment:
- iTEVA Security
Attachment(s):
| File | Last Modified |
|---|---|
| iTEVA Security Software Installation Manual.pdf | October 05, 2023 |