"INF" error in Qtegra
Issue
"INF" error in Qtegra
Environment
- Qtegra
- ECCN:EAR99
Resolution
“INF” here means “Infinity”, so basically there’s a calculation that is resulting in a very large number that is outside the range of possible values for that number type. Is there some form of interference correction set? This shouldn’t normally happen unless something is configured badly.
Analysis
Cause