Thermo Fisher Scientific
Exhaust Error
Issue
Plasma wouldn't light due to incorrect exhaust flow
Environment
  • iCAP Q
  • iCAP RQ
  • ECCN: EAR99
Resolution
Viewed the plasma exhaust reading in the plasma tab of instrument control.
It was reading 1.02mbar. Adjusted damper for exhaust to bring down flow to 0.45mbar.
Analysis
Cause
Exhaust too high, due to exhaust fans being replaced and reset to highest flow.
  • Was this article helpful?