How to Replace the Spray Chamber
This video will show you how to replace the spray chamber on an iCAP Q / RQ / TQ. The spray chamber may need to be replaced or checked for tightness when troubleshooting issues for stability verification failures, high oxides, etc.
Description:
This video will show you how to replace the spray chamber on an iCAP Q / RQ / TQ. The spray chamber may need to be replaced or checked for tightness when troubleshooting issues for stability verification failures, high oxides, etc.
Environment:
- iCAP Q
- iCAP RQ
- iCAP TQ
- ECCN: EAR99