Thermo Fisher Scientific
Mass Calibration Verification Failure, iCAP Qnova Series
Issue
Mass Calibration Verification Failure for Peak Width or Offset in Performance Report
Environment
  • iCAP Q
  • iCAP RQ
Resolution
Perform a mass calibration using the mass calibration wizard in the Qtegra software. The mass calibration will need to be performed in STD (or STDS mode if using High Sensitivity interface) using the Setup or Calibration (i.e.P/N Thermo-5A) solution.
Analysis
Cause
  • Was this article helpful?