Thermo Fisher Scientific
Performance report not showing result.
Issue
Not all chosen stability masses seen in the performance report.
Environment
  • iCAP Q
  • iCAP Qnova
  • Qtegra
Resolution
  1. Open file manager and select Performance ReportStep 1.png
  2. Click the following (arrows) and refresh
    Step 2.png
  3. Results should become visible
    Step 3.png
Cause
This can happen if you add elements to be checked in the performance report.
  • Was this article helpful?