Plasma going out
Issue
Plasma flow settings get changed to zero
Environment
- iCAP Q
- iCAP RQ
- Qtegra
- ECCN: EAR99
Resolution
- Check the post ignition settings to insure no zero gas flows are set for torch coolant, auxiliary or nebulizer gas.
- The last tune setting state may have been saved with all the gas flows set to zero.
- Right click and set an older state to the post-ignition default settings; or
- Go to “Edit” and double-click on a state that was successful in the past. Select Apply Tune Settings so that this state moves to top of the list.