Thermo Fisher Scientific
Replacing the Tensioners on Mini Peristaltic Pump
Issue
Tensioners on Peristaltic Pump Need Replaced
Environment
  • iCAP Q
  • iCAP Qnova
Resolution
Analysis
Sample and/or internal standard solution is not flowing smoothly even after optimizing the tension on the pump tubing. It is possible that the tensioner(s) is bent and needs to be replaced. 
Cause
  • Was this article helpful?