Thermo Fisher Scientific
Bürkert Gas Module - Field Upgrade Manual
Description: 
If the instrument is running with a Qtegra version lower than 2.4 SP1, it must be upgraded before the
hardware modification, owing to an incompatibility with the firmware.
Environment:
Bürkert
  • Was this article helpful?