Thermo Fisher Scientific
iCAP 7000 - Sorting Analystes Alphabetically in a Report within Qtegra for ICP-OES
Description: 
While in the LabBook navigate to Reports and select the report in which you would like to have the analytes sorted alphabetically. In this example, the sorting of anlaytes alphabetically will be demonstrated using the Concentrations report
Environment:
iCAP 7000
Qtegra