Thermo Fisher Scientific
Adding Elements, Molecules, and Ratios to the Data Display in Instrument Control
Issue
  • Data Display elements and ratios are no longer present 
  • Would like to add a new element, molecular or ratio to the Data Display
Environment
  • iCAP Q
  • iCAP Qnova Series
Resolution
1) Click on the Analytes tab
Analytes.PNG
2) Click on Elements tab
Elements tab.PNG
3) Select the elements which you would like to add to the Data Display by clicking on the respective element in the periodic table. You can right-click on an element to select a specific isotope
Below is a list of elements to add in respect to monitoring performance according to the elements contained within a standard Tune B solution
 Tune B Analytes.PNG
4) Click on Molecules tab
Molecules tab.PNG
5) Select Ce.O and then select 140Ce.16O by placing a tick in the checkbox next to it. Complete a similar step for Ba++ to add 137Ba++ and Cl.O to add 35Cl.16O. These molecules need to be added in order to monitor the oxide, doubly charged and KED sensitivity factor.
Selecting Molecules.PNG
Select Bkg as well and add Bkg220.7 and Bkg4.5 in order to monitor background masses
Selecting Bkg.PNG
6) Change the dwell time to 0.1s for 137Ba++, CeO, ClO and background masses in order to get a more stable signal
 changing Dwell Times.PNG
7) Click on Formula Table tab and then click on AnalyteFunction button three times to add three rows of formulas
Formula table.PNG
8) Set up the formulas similar to that in the following figure. Having the Operator set to '%' will automatically convert the Oxide and Doubly charged ratio to a percentage