OXSAS Version 2.x Update
Description:
OXSAS_2.7.1 can now be downloaded here: https://we.tl/t-0VIvNEAXiE
OXSAS installer will take care to update the SQL server (if needed), DO NOT INSTALL SQL SERVER MANUALLY.
For those having SQL server 2012 already installed, OXSAS_2.7.1 without SQL installer can now be downloaded here: https://we.tl/t-30RzovCzN9
Do not use this link to update OXSAS from version 2.3 or older versions (please use the previous link instead).
Contains:
Setup_Oxsas_2.7.x.yyyy.exe contains only OXSAS software installer (slightly faster to download - it does not contains SQL server)
Abbreviated procedure:
- Download the executable file by clicking on OXSAS_Update_2.m.s.b.exe" file in the right frame (attachments list).
- Run the file on the OXSAS computer and follow on-screen instructions. This will remove the previous version and install the new (update) OXSAS version.
- Start OXSAS. If prompted for updating the databases version, click "Yes" button. Repeat the same for the other databases (ANALYSES, ARCHIVES, LngMgtOXSAS).
Notes:
-
You must be logged with Computer Administrator level in Windows to perform the OXSAS version update.
-
The update of the databases (if required) will take several minutes.DO NOT INTERRUPT THAT PROCESS.
-
Warning: if the installed version is below 2.1.1a, the new update kit may ask you to reboot sometime in the update process.
-
DO NOT REBOOT when prompted for; simply let the update kit run to the end (“Finish” button) then reboot.
-
Alternatively, you may uninstall OXSAS from the Control Panel before installing the update.
Environment:
- OXSAS version 2.x (this update can be uploaded on existing OXSAS version 2.m.s only)
- Windows 10 is supported since OXSAS 2.5.1
- SQL server 2012 is supported since OXSAS 2.4.1
Attachment(s):
| File | Last Modified |
|---|---|
| 309884-A_EN [OXSAS 2.7.1.,Release Notes] 0821.pdf | January 06, 2022 |