Thermo Fisher Scientific
Total Materia Software Update
Description: 
The latest version of Total Materia is now available and must be used with OXSAS 2.8.1.
Please note that the previous version of Total Materia is no longer compatible with OXSAS 2.8.1.
🔹 Dongle Requirement: This version still requires the original dongle for access—customers will need to update the software but continue using their existing dongle.
🔹 New Instruments: All new instruments will be shipped with this updated version pre-installed from the factory.
Environment:
  • OXSAS version 2.8.1
  • Was this article helpful?