UQED Software Update
Description:
The UniQuant version upgrade kit is attached.
Environment:
- UniQuant version 5 is for WD-XRF and Win OS up to Windows 10
- UniQuant version 6 and 7 is for ED-XRF and Win OS up to Windows 10
- The integrated version of UniQuant, UQi, as a module of OXSAS is for WD-XRF only.
Attachment(s):
| File | Last Modified |
|---|---|
| UniQuant 6.37.zip | April 14, 2026 |
| UniQuant 6.30.zip | February 14, 2025 |