Thermo Fisher Scientific
Thermo Scientific AI/AS 1310 Liquid Autosampler Specifications
The Thermo Scientific AI/AS 1310 Series autosampler offers advanced sampling automation for gas chromatography liquid injections, with systems ranging from an 8-position AI 1310 to a 155-sample capacity AS 1310. Features include syringe thermal stability, cross-contamination-free rinsing, easy system setup, RoHS compliance, and system upgradability. The AS 1310 Gemini kit enables simultaneous injections on two channels. The system supports various syringes and injection parameters.
Description: 
Thermo Scientific AIAS 1310 Liquid Autosampler Specifications
Environment:
AIAS1310
  • Was this article helpful?