PS52260_E - TRACE 1300 Series Gas Chromatograph Specification
The Thermo Scientific TRACE 1300 Series Gas Chromatograph offers advanced performance for QA/QC and routine labs, featuring user-exchangeable, instant connect injectors and detectors, enhancing flexibility and reducing ownership costs. The TRACE 1310 includes a touch-screen interface for method development labs, while the TRACE 1300 is ideal for basic labs with its single-button system. Both models ensure high productivity with features like the Helium Saver Module and fast oven performance.
Description:
Product Specification Sheet
Environment:
- TRACE 1300 GC
- TRACE 1310 GC
Attachment(s):
| File | Last Modified |
|---|---|
| PS52260_E - TRACE 1300 Series Gas Chromatograph Specification.pdf | November 11, 2021 |