Thermo Fisher Scientific
Alignment errors for sample trays and centering plates
For AI/AS 1310 and 3000 experiencing alignment errors, ensure the trays for the AS 1310 sit flush on the mount, aligning the hub grooves and tray tabs properly to maintain the correct height. Improper alignment will prevent the sample arm from picking up samples. Also, align the SSL centering plate openings correctly to avoid the autosampler assuming the inlet is a PTV, which can cause errors. Refer to figures for proper alignment details.
Issue
AI/AS 1310 and 3000 experiencing alignment errors for sample trays and centering plates.
Environment
  • AS 1310
  • AI 1310
Resolution
  • The trays for the AS 1310 must sit flush on the mount as shown in figures 1 and 2.
Note: The distance between the arrows. Figures 3 and 4 show improper alignment.
Note: The gap between the hub and tray. The Hub grooves and the Tray tabs must be aligned to allow the tray to fit securely on the hub and provide the proper height for the tray.
  • If this height is not correct the sample arm will not pick up the samples.
  • See Figure 5 for a picture of the grooves on the hub and and Figure 6 for picture of tabs on the sample tray.  
  • See Figure 7 for the alignment of the SSL centering plate openings.
  • If the opening on the metal plate is not aligned with the half moon cutout of the plastic centering plate, the autosampler assumes the inlet is a PTV and will not go completely down on the SSL.
  • This may cause an error if the alignment is not correct.  
clipboard_e9a39140fd3a8a24bcde08ebde2d20684.png
  • Was this article helpful?