Thermo Fisher Scientific
TriPlus Liquid Syringe Tool Replacement
Topic
TriPlus—LS Syringe Replacement
Environment
TriPlus RSH SMART
Steps/Procedure
  1. Remove the retaining nut from the Liquid Syringe Tool.
  2. Push the needle out from the bottom.
  3. Gently pull the needle out of the tool.
  4. Then insert the syringe such that the flat side of the flange matches with the cut-out of the upper part of the holder.
  5. Screw on the retaining nut.
Additional Information
Make sure to adhere to the safety rules as indicated in the TriPlus RSH SMART user manuals.