TriPlus RSH Preventive Maintenance Intervals
Topic
Here is the maintenance schedule for the TriPlus RSH autosampler.
Environment
- TriPlus RSH
- SMART or RSH Classic
Maintenance
| Maintenance Step | Interval |
| Clean exterior surfaces using lint-free cloth and soapy water | Quarterly or as needed |
| Lubricate RSH Z-Head | Every 6 months or yearly |
| Manual cleaning, replacement of Syringe Plunger, fibers, and ITEX traps | Metal Plunger: Remove weekly and clean with an alcohol-soaked wipe |
| Gas-tight Syringe (polymer tip) | Depending on the application clean regularly or replace the plunger |
| Dispose of wash or waste solvent reservoirs | Quarterly or as Needed |
| Replace septum | After 100 + injections or as needed to avoid contamination |
| Check alignment of module positions | Biweekly or quarterly depending on run frequency |
Additional Information
The major maintenance for the TriPlus usually involves replacing consumables such as the fibers, syringes, and ITEX traps.