TriPlus RSH SMART Autosampler Sampling Techniques Overview
Topic
The TriPlus RSH SMART autosampler provides automated sample preparation and introduction for GC and GC-MS analysis.
The training covers three primary sampling technique groups:
-
Static Headspace (SHS)
-
Solid Phase Microextraction (SPME), including SPME Arrow
-
In-Tube Extraction (ITEX)
The liquid sampling tool is not covered because the source training describes the liquid sampling concept as straightforward.
Environment
-
Thermo Scientific TriPlus RSH SMART autosampler
-
GC or GC-MS analysis
-
Sampling tools discussed in the training:
-
Static Headspace
-
SPME
-
SPME Arrow
-
ITEX
-
Steps/Procedure
-
Select the sampling technique appropriate for the analytical application.
-
Use Static Headspace when volatile compounds need to be partitioned from a gaseous, liquid, or solid sample matrix into the headspace.
-
Use SPME when analytes need to be extracted onto a coated fiber before thermal desorption into the GC.
-
Use SPME Arrow when the application benefits from the larger sorption volume and surface area of the Arrow fiber design.
-
Use ITEX when volatile compounds need to be trapped and concentrated using a sorbent-containing micro-trap.
-
Configure the selected sampling technique within the applicable instrument method.
Additional Information
The training identifies the following benefits of the TriPlus RSH SMART autosampler:
-
Rapid automation for GC and GC-MS sample analysis
-
Robustness and reliability
-
Flexibility through multiple sampling tools
-
Reduced downtime
-
Increased productivity and throughput