Thermo Fisher Scientific
TriPlus SPME Fiber Installation
Topic
TriPlus—SPME Fiber Replacement
Environment
TriPlus RSH SMART
Steps/Procedure
  1. Unscrew the cap from the SPME tool adapter.
  2. Push the black fiber holder all the way in until the end shows.
  3. Screw the SPME fiber needle into the fiber holder.
  4. Then pull the holder back and recap the cap on the adapter.
  5. Insert the SPME tool adapter into the tool and align its position as demonstrated here.
  6. Cap and screw down the retaining nut.
Additional Information
Make sure to adhere to the safety rules as indicated in the TriPlus RSH smart user manuals.