TriPlus Tool Position Verification
Topic
Maintenance—Checking Calibration
Environment
TriPlus RSH
Steps/Procedure
- Here is the handheld terminal also known as the “Gameboy.”
- Using the dial, scroll to the tool desired, and press the button in the middle to enter.
- Press the left button to select “Options.”
- Scroll and select “Check Tool Calibration.”
- Press the right button to select “Check.”
- The X, Y, Z head will move to the saved position.
- Visually inspect to ensure correct positioning.
- When check is satisfactory, click “Next,” then click “Ok” to complete.
- Click back to return to the home page and return X, Y, Z robotic head to its calibrated home position.
Additional Information
Make sure to adhere to the safety rules as indicated in the TriPlus RSH user manuals.