TRACE 1300 GC Series Module Calibration Software V 1.00.73
The Trace 1300 Series Calibration Software, for service team use, allows module calibration with a flowmeter, thermometer, and barometer. Installation involves running "setup.exe" and configuring the GC system via IP address. Calibration starts by recognizing instrument configuration, selecting parameters, and following step-by-step instructions. Oven temperature and electronic cassette serial numbers can also be set. Images illustrate steps and final reports.
Description:
Updated version of the TRACE 1300 Series Calibration Software
Environment:
- TRACE 1300 GC Series Calibration Software v 1.00.73
- For Field Calibration of Modules and Oven
Attachment(s):
| File | Last Modified |
|---|---|
| TRACE 1300 GC Series Calibration Software 1.00.73.zip | November 12, 2021 |
| Rev A - TRACE 1300 GC Series Calibration Software 1.00.73 Installation Instructions.pdf | November 12, 2021 |