Thermo Integrion Agent Quick Start Guide
The Thermo Integrion Agent Quick Start Guide outlines the steps to install and configure the Thermo Integrion agent data service. The process includes pre-installation tasks, installation tasks, and post-installation tasks. Ensure Chromeleon CM7.2.SR4 or higher is installed, complete the installation using the ThermoInstaller CD, and verify the setup.
Description:
This Quick Start Guide describes the steps to install and configure the Thermo Integrion agent data service and other supplementary components.
Environment:
- Integrion HPIC System Instrument PC
- Chromeleon CM7.2.SR4 (build 8179) or higher
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Integrion Agent Quick Start Guide.pdf | November 21, 2021 |