031917 - Rev 02 - ICS-1500 Ion Chromatography System
The document is the operator’s manual for the Dionex ICS-1500 Ion Chromatography System, detailing its operation, maintenance, and troubleshooting. It covers system components, setup procedures, and safety information. The manual includes instructions for system installation, eluent preparation, sample processing, and routine maintenance. Troubleshooting guides and service procedures are provided for common issues.
Description:
The Dionex ICS-1500 Ion Chromatography System ICS-1500 performs ion analyses using suppressed or non-suppressed conductivity detection. An ion chromatography system typically consists of a liquid eluent, a high-pressure pump, a sample injector, a guard and separator column, a chemical suppressor, a conductivity cell, and a data collection system.
Environment:
- The Dionex ICS-1500 Ion Chromatography System
Attachment(s):
| File | Last Modified |
|---|---|
| 031917 - Rev 02 - ICS-1500 Ion Chromatography System.PDF | December 15, 2021 |