065289 - Rev 03 - Dionex ICS-1100 Ion Chromatography
The Thermo Scientific Dionex ICS-1100 Operator Manual provides detailed guidance on operating, maintaining, and troubleshooting. The system conducts ion analyses using conductivity detection and involves stages like eluent delivery, sample injection, separation, suppression, detection, and data analysis. It describes system features, components, and the Chromeleon software interface. The manual also includes safety information, error messages, and maintenance procedures for optimal performance.
Description:
The Thermo Scientific Dionex™ ICS-1100 Ion Chromatography System (Dionex ICS-1100) performs ion analyses using suppressed or non-suppressed conductivity detection. An ion chromatography system typically consists of a liquid eluent, a high-pressure pump, a sample injector, a guard and separator column, a chemical suppressor, a conductivity cell, and a data collection system.
Environment:
Thermo Scientific Dionex™ ICS-1100 Ion Chromatography System (Dionex ICS-1100)
Attachment(s):
| File | Last Modified |
|---|---|
| 065289 - Rev 03 - Dionex ICS-1100 Ion Chromatography.PDF | December 14, 2021 |