065291- Rev 03 - ICS-2100 - Dionex ICS-2100 Ion Chromatography System Operators Manual
The Dionex ICS-2100 Ion Chromatography System Operator’s Manual details operation, maintenance, and troubleshooting for the ICS-2100. It describes ion analysis stages, system features, components, and the Chromeleon software interface used for control.
Description:
The Thermo Scientific Dionex ICS-2100 Ion Chromatography System Dionex ICS-2100 performs ion analyses using suppressed or non-suppressed conductivity detection. An ion chromatography system typically consists of a liquid eluent, a high-pressure pump, a sample injector, a guard and separator column, a chemical suppressor, a conductivity cell, and a data collection system.
Environment:
- Thermo Scientific Dionex ICS-2100 Ion Chromatography System
Attachment(s):
| File | Last Modified |
|---|---|
| 065291- Rev 03 - ICS-2100 - Dionex ICS-2100 Ion Chromatography System Operators Manual.PDF | October 09, 2025 |