How to Activate a New Eluent Generator Cartridge
To activate a new eluent generator cartridge in Chromeleon 7 for ICS2000/2100 and 3000, ensure the cartridge cable is connected, the IC system is powered on, and the cartridge serial number is entered in the Instrument Configuration Manager. Open Chromeleon 7 Console, access the Instruments Category Bar, and follow the appropriate steps to select and activate the cartridge, ensuring the mode is set to "On" and verifying there are no errors in the Instrument Audit Trail.
Description:
The attached document describes how to activate a new eluent generator cartridge.
Environment:
- Chromeleon 7
- ICS-2000
- ICS-2100
- ICS-3000
Attachment(s):
| File | Last Modified |
|---|---|
| How to Activate a New Eluent Generator Cartridge.PDF | January 09, 2022 |